Learn More
Selleck Chemical LLC Zegocractin CM 4620-5mg
Zegocractin (CM 4620) is an inhibitor of calcium-release activated calcium-channel (CRAC channel). CM4620 inhibits Orai1/STIM1-mediated Ca2+ currents and Orai2/STIM1-mediated currents with IC50 of near 120 nM and 900 nM, respectively.