Learn More
ZeptoMetrix Immuno-Tek Goat IgG ELISA Kit
Shop All ZeptoMetrix Corporation Products

Includes
One 96 well microplate, 12mL detector antibody, 7mL of Goat IgG standard, 100mL assay diluent, 125mL of plate wash buffer, 12mL substrate, 12mL stop solution, 1 pack of microplate sealers, 1 plastic bag.
- Contains ready–to–use reagents
- Takes less than two hours to perform
- Microplate and detector antibody reacts with all subclasses of goat IgG
Specifications
Product Type
Immuno-Tek Goat IgG ELISA kit
Sufficient For
96-well Microplate
For Use With (Application)
Designed for the measurement of Goat IgG in Goat colostrum, milk, Serum, Plasma or other biological fluids, the microplate and Detecto r Antibody in the kit have been specifically balanced to react uniformly with all subclasses of Goat IgG
Kit Contents
1 96-well microplate, 1 pack of microplate sealers, 1 plastic bag, 100 mL assay diluent, 12 mL detector antibody, 12 mL stop solution, 12 mL substrate, 125 mL plate wash buffer, 7 mL of Goat IgG standard
Quantity
96 Determinations
Storage Requirements
2°C to 8°C
Research purposes only. Not for in vitro diagnostic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.