Learn More
HANNA INSTRUMENTS INC Zero Oxygen Solution Set (500 mL)
SDP
Supplier:  HANNA INSTRUMENTS INC HI7040L
Encompass
HI7040L is a high quality, bicomponent zero oxygen solution made from reagent grade chemicals. Hanna dissolved oxygen solutions have the lot number and expiration date clearly marked on the label and are air tight with a tamper-proof seal to ensure the quality of the solution.Prepared with reagent grade chemicalsHassle-free, easy to mix chemical component bottlesAir-tight seal to ensure the quality of the solution
Catalog No. NC1805035
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.