Learn More
Cole-Parmer Oakton Zero Oxygen Calibration Solution, 500 mL
Supplier:  Cole-Parmer 0065300
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Chemical oxygen scrubbers remove oxygen from a solution to provide fast calibration results. It is manufactured, according to the APHA and ASTM formulation, using high-purity, oxygen-free water.
Catalog No. NC9964632
missing translation for 'QuantityLeft'