Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zero Oxygen Standard for Calibration of Dissolved Oxygen Meters, Reagents
Click to view available options
Quantity:
120 mL
500 mL
Specifications
Specifications
Chemical Name or Material | Zero Oxygen Standard for Calibration of Dissolved Oxygen Meters |
Packaging | Poly Bottle |
CAS | 7732-18-5, 7757-83-7 |
Quantity | 120 mL |
Color | Colorless |
Grade | Reagent |
Boiling Point | 101°C |
Specific Gravity | 1.0311 |
Solubility | Completely miscible |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction