Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zileuton, Tocris Bioscience™

Catalog No. p-4784852 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

5-LOX inhibitor; orally bioavailable

Orally active 5-lipoxygenase (5-LOX) inhibitor that inhibits LTB4 synthesis (IC50 values are 0.56, 2.3 and 2.6 μM in dog, rat and human blood respectively). Inhibits antigen-induced contraction of tracheal strips in vitro (IC50 = 6 μM) and exhibits antiasthmatic activity in vivo. Also weakly inhibits CYP1A2 (Ki = 66 - 98 μM).

Chemical Identifiers

CAS 111406-87-2
Molecular Formula C11H12N2O2S
Molecular Weight (g/mol) 236.29
MDL Number MFCD00866097
InChI Key MWLSOWXNZPKENC-UHFFFAOYNA-N
Synonym zileuton, zyflo, leutrol, zileutonum, 1-1-benzo b thiophen-2-yl ethyl-1-hydroxyurea, zyflo cr, zileutonum inn-latin, abbott 64077, n-1-benzo b thien-2-ylethyl-n-hydroxyurea, zyflo tn
PubChem CID 60490
ChEBI CHEBI:10112
IUPAC Name 1-[1-(1-benzothiophen-2-yl)ethyl]-1-hydroxyurea
SMILES CC(N(O)C(N)=O)C1=CC2=CC=CC=C2S1

Specifications

Quantity 10 mg
Formula Weight 236.29
Percent Purity >99%
Chemical Name or Material Zileuton
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.