Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Zinc-Cast Slotted Masses Set

Alternate for expensive brass weights.
Supplier: Eisco™ PHZNW100SET
Description
- Zinc die-casted with powder coat
- Set of nine weights and one hanger
- Accuracy ±2%
Specifications
Powder-coated Slotted Mass Set | |
Zinc Die-cast | |
100 g | |
Set of 9 x 10 g weights and 1 x 10 g hanger |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction