Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Zinc Chloride
SCI_ED

Catalog No. p-4416492 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7646-85-7
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
PubChem CID 5727
ChEBI CHEBI:49976
IUPAC Name dichlorozinc
SMILES Cl[Zn]Cl

Specifications

Color White
Physical Form Solid
Quantity 500 g
Assay Percent Range >95%
Linear Formula ZnCl2
Formula Weight 136.29
Grade Reagent
Packaging Reagent Grade
Chemical Name or Material Zinc Chloride
ShelfLife 1 year
DOTInformation DOT Class 8, III, : Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.