Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Dimethyldithiocarbamate 97.0+%, TCI America™
SDP

Catalog No. p-7126392 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 137-30-4
Molecular Formula C6H12N2S4Zn
Molecular Weight (g/mol) 305.796
MDL Number MFCD00064797
InChI Key DUBNHZYBDBBJHD-UHFFFAOYSA-L
Synonym ziram, zinc dimethyldithiocarbamate, carbazinc, fuclasin, methazate, zarlate, aazira, cymate, tsimat, zinc bis dimethyldithiocarbamate
PubChem CID 8722
ChEBI CHEBI:79736
IUPAC Name zinc;N,N-dimethylcarbamodithioate
SMILES CN(C)C(=S)[S-].CN(C)C(=S)[S-].[Zn+2]

Specifications

Melting Point 257°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 305.80
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Zinc Dimethyldithiocarbamate
EINECSNumber (2)-1833&(9)-0607
RTECSNumber ZH0525000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.