Learn More
Zinc, granular, 20 mesh, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Gray |
Melting Point | 420.0°C |
Boiling Point | 907.0°C |
Density | 7.1400g/mL |
Linear Formula | Zn |
Quantity | 500 g |
Fieser | 01,1276; 02,459; 03,334; 04,574; 05,753; 06,672; 07,426; 08,532; 10,459; 11,98; 12,568; 13,298; 14,111; 16,106; 17,406 |
Formula Weight | 65.38 |
Percent Purity | ≥99.7% (Complexometry) |
Packaging | Plastic bottle |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.