Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Zinc, Mossy
SCI_ED

Catalog No. p-4488665 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Lab Grade
Reagent Grade
This item is not returnable. View return policy
This item is not returnable. View return policy

Reacts with water or moisture

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7440-6-6
Synonym Zinc Dust

Specifications

Color Blue to White
Density 7.14g/mL
Assay Percent Range 100% w/w
Linear Formula Zn
Quantity 500 g
Formula Weight 65.38
Packaging Lab Grade
Chemical Name or Material Zinc, Mossy
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.