Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Zinc, Sheet
SCI_ED

Catalog No. p-4416756 Shop All Fisherbrand Products
Click to view available options
Quantity:
6 x 12 in.
12 x 12 in.
Packaging:
Solid
This item is not returnable. View return policy
This item is not returnable. View return policy

Reacts with water or moisture

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7440-6-6

Specifications

Color Gray
Density 7.14g/mL
Assay Percent Range 100% w/w
Linear Formula Zn
Quantity 6 x 12 in.
Synonym Zinc Dust
Formula Weight 65.38
Packaging Solid
Chemical Name or Material Zinc, Sheet
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.