Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zincon Sodium Salt, MP Biomedicals™

Catalog No. p-4679581 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 62625-22-3
Molecular Formula C20H16N4NaO6S
Molecular Weight (g/mol) 463.42
InChI Key IABRINWJUBAIIE-JJECXDOKSA-N
Synonym 2-Carboxy-2'-hydroxy-5'-sulfoformazylbenzene
PubChem CID 131856391
IUPAC Name 2-[2-[(Z)-N-[(Z)-(6-oxo-3-sulfocyclohexa-2,4-dien-1-ylidene)amino]-C-phenylcarbonimidoyl]hydrazinyl]benzoic acid;sodium
SMILES C1=CC=C(C=C1)C(=NN=C2C=C(C=CC2=O)S(=O)(=O)O)NNC3=CC=CC=C3C(=O)O.[Na]

Specifications

Quantity 1 g
Formula Weight 462.4
Physical Form Solid
Chemical Name or Material Zincon Sodium Salt
EINECSNumber 263-651-1
Recommended Storage Store at 2° to 8°C.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.