Learn More
Zirconium powder, APS 2-3 micron, packaged in water, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged in water
Specifications
Specifications
Color | Gray |
Quantity | 25 g |
UN Number | UN1358 |
Odor | Odorless |
Particle Size | APS 2 to 3 Micron |
Form | Powder |
Chemical Name or Material | Zirconium powder |
Safety and Handling
Hazard Category | H228 |
Hazard Statement | GHS H Statement H228 Flammable solid. |
Precautionary Statement | P210-P240-P241-P280-P370+P378q |
DOTInformation | Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: ZIRCONIUM POWDER, WETTED |
EINECSNumber | 231-176-9 |
RTECSNumber | ZH7070000 |
TSCA | Yes |
storageNote1 |
|
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.