Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Zofenopril calcium

Catalog No. p-7214722
Click to view available options
Quantity:
100 mg
500 mg

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 81938-43-4
Molecular Formula 0·5 Ca

Specifications

Quantity 100 mg
Solubility Solubility aq. soln.: .. Other solubilities: soluble in dmso, insoluble in methanol
Melting Point 249.0°C to 251.0°C
Formula Weight 448.58
InChI Key NSYUKKYYVFVMST-LETVYOFWSA-L
SMILES [Ca++].C[C@H](CSC(=O)C1=CC=CC=C1)C(=O)N1C[C@H](C[C@H]1C([O-])=O)SC1=CC=CC=C1.C[C@H](CSC(=O)C1=CC=CC=C1)C(=O)N1C[C@H](C[C@H]1C([O-])=O)SC1=CC=CC=C1
IUPAC Name calcium bis((2S,4S)-1-[(2S)-3-(benzoylsulfanyl)-2-methylpropanoyl]-4-(phenylsulfanyl)pyrrolidine-2-carboxylate)
Molecular Weight (g/mol) 897.16
Chemical Name or Material Zofenopril calcium
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Very toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement Avoid release to the environment.

Collect spillage.

Dispose of contents/container to an approved waste disposal plant.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.