Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zolmitriptan, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent 5-HT1B/1D/1F agonist (Ki values are 0.63, 5.01 and 63.09 nM for h5-HT1D, h5-HT1B and h5-HT1F receptors respectively). Inhibits trigeminal nerve stimulation-mediated neurogenic plasma protein extravasation in guinea pigs; blocks long excitatory postsynaptic potentials (EPSPs) in a rat model of spinal cord injury.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 287.36 |
Percent Purity | >99% |
Chemical Name or Material | Zolmitriptan |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction