Learn More
Medchemexpress LLC HY-B0124 200mg Medchemexpress, Zonisamide CAS:68291-97-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0124200MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-B0124 200mg Zonisamide CAS:68291-97-4 Zonisamide (AD 810; CI 912) is an inhibitor of zinc enzyme carbonic anhydrase (CA), with Kis of 35.2 nM and 20.6 nM for human mitochondrial isozyme hCA II and hCA V, respectively. Zonisamide has antiepileptic activity. Zonisamide can be used for the rsearch for epilepsy, seizures and Parkinson's disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2370
missing translation for 'QuantityLeft'