Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zonisamide, Tocris Bioscience™

Catalog No. p-4784876 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Blocks voltage-sensitive Na+ and Ca2+ channels (T-type)

Antiepileptic that possesses a broad spectrum anticonvulsant and mechanistic profile. Blocks voltage-sensitive Na+ and T-type Ca2+ channels, stimulates BKCa channels, modulates GABA, glutamate and monoamine neurotransmission, inhibits lipid peroxidation and scavenges hydroxyl and nitric oxide free radicals. Displays neuroprotective and antiParkinsonian activity.

Chemical Identifiers

CAS 68291-97-4
Molecular Formula C8H8N2O3S
Molecular Weight (g/mol) 212.22
MDL Number MFCD00865316
InChI Key UBQNRHZMVUUOMG-UHFFFAOYSA-N
Synonym zonisamide, zonegran, exceglan, excegram, 1,2-benzisoxazole-3-methanesulfonamide, zonisamida, zonisamidum, zonisamidum latin, zonisamida spanish, excegran
PubChem CID 5734
ChEBI CHEBI:10127
IUPAC Name (1,2-benzoxazol-3-yl)methanesulfonamide
SMILES NS(=O)(=O)CC1=NOC2=CC=CC=C12

Specifications

Quantity 10 mg
Formula Weight 212.22
Percent Purity >99%
Chemical Name or Material Zonisamide
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.