Learn More
Zymo Research Corporation Zyppy™ Plasmid Miniprep Kit, 800 Preps
Supplier: Zymo Research Corporation D4037
The Zyppy™ Plasmid Miniprep Kit features a pellet-free modified alkaline lysis method that bypasses bacterial culture centrifugation and resuspension steps common to classical plasmid preparation procedures. Simply add the uniquely formulated 7X Lysis Buffer directly to your bacterial culture, neutralize, and then purify using the provided unique column technology. Additionally, the innovative colored buffers included in the kit permit error-free visualization and identification of complete bacterial cell lysis and neutralization. The Zyppy™ Plasmid Miniprep Kit is the fastest and easiest method available to separate plasmid DNA from E. Coli efficiently. The plasmid DNA is of the highest quality, endotoxin-free, and is well suited for use in transfection, bacterial transformation, restriction endonuclease digestion, DNA ligation, PCR, transcription, sequencing, and other sensitive downstream applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.