Assay Details
Gene Symbol: LOC5572804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846995.1 | XP_021702687.1 | 2 - 3 | 1048 | 56 | |
XM_021846997.1 | XP_021702689.1 | 2 - 3 | 1048 | 56 | ||
XM_021846998.1 | XP_021702690.1 | 2 - 3 | 1309 | 56 | ||
XM_021846999.1 | XP_021702691.1 | 2 - 3 | 1003 | 56 | ||
XM_021847000.1 | XP_021702692.1 | 2 - 3 | 1486 | 56 | ||
XM_021847001.1 | XP_021702693.1 | 2 - 3 | 919 | 56 | ||
XM_021847002.1 | XP_021702694.1 | 2 - 3 | 1052 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
