Assay Details
Gene Symbol: LOC5572804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846995.1 | XP_021702687.1 | 6 - 7 | 3317 | 99 | |
XM_021846997.1 | XP_021702689.1 | 6 - 7 | 3317 | 99 | ||
XM_021846998.1 | XP_021702690.1 | 6 - 7 | 3578 | 99 | ||
XM_021846999.1 | XP_021702691.1 | 6 - 7 | 3272 | 99 | ||
XM_021847000.1 | XP_021702692.1 | 6 - 7 | 3755 | 99 | ||
XM_021847001.1 | XP_021702693.1 | 6 - 7 | 3188 | 99 | ||
XM_021847002.1 | XP_021702694.1 | 6 - 7 | 3321 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
