Assay Details
Gene Symbol: LOC5568181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848214.1 | XP_021703906.1 | 5 - 6 | 1062 | 111 | |
XM_021848215.1 | XP_021703907.1 | 4 - 5 | 1412 | 111 | ||
XM_021848216.1 | XP_021703908.1 | 5 - 6 | 1344 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
