Assay Details
Gene Symbol: LOC5568181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848214.1 | XP_021703906.1 | 6 - 7 | 1219 | 73 | |
XM_021848215.1 | XP_021703907.1 | 5 - 6 | 1569 | 73 | ||
XM_021848216.1 | XP_021703908.1 | 6 - 7 | 1501 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
