Assay Details
Gene Symbol: LOC5568181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848214.1 | XP_021703906.1 | 9 - 10 | 2322 | 103 | |
XM_021848215.1 | XP_021703907.1 | 8 - 9 | 2672 | 103 | ||
XM_021848216.1 | XP_021703908.1 | 9 - 10 | 2604 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
