Assay Details
Gene Symbol: LOC5579652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021853767.1 | XP_021709459.1 | 5 - 6 | 3076 | 69 | |
XM_021853768.1 | XP_021709460.1 | 5 - 6 | 3103 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
