Assay Details
Gene Symbol: LOC5572977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842723.1 | XP_021698415.1 | 10 - 11 | 7935 | 64 | |
XM_021842724.1 | XP_021698416.1 | 11 - 12 | 8545 | 64 | ||
XM_021842725.1 | XP_021698417.1 | 11 - 12 | 8541 | 64 | ||
XM_021842726.1 | XP_021698418.1 | 11 - 12 | 8519 | 64 | ||
XM_021842727.1 | XP_021698419.1 | 11 - 12 | 8515 | 64 | ||
XM_021842728.1 | XP_021698420.1 | 10 - 11 | 8395 | 64 | ||
XM_021842729.1 | XP_021698421.1 | 9 - 10 | 7770 | 64 | ||
XM_021842730.1 | XP_021698422.1 | 8 - 9 | 3511 | 64 | ||
XM_021842731.1 | XP_021698423.1 | 8 - 9 | 3303 | 64 | ||
XM_021842732.1 | XP_021698424.1 | 7 - 8 | 3105 | 64 | ||
XM_021842733.1 | XP_021698425.1 | 10 - 11 | 8395 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
