Assay Details
Gene Symbol: LOC5572977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842723.1 | XP_021698415.1 | 4 - 5 | 5442 | 115 | |
XM_021842724.1 | XP_021698416.1 | 5 - 6 | 6052 | 115 | ||
XM_021842725.1 | XP_021698417.1 | 5 - 6 | 6048 | 115 | ||
XM_021842726.1 | XP_021698418.1 | 5 - 6 | 6026 | 115 | ||
XM_021842727.1 | XP_021698419.1 | 5 - 6 | 6022 | 115 | ||
XM_021842728.1 | XP_021698420.1 | 4 - 5 | 5902 | 115 | ||
XM_021842729.1 | XP_021698421.1 | 4 - 5 | 5901 | 115 | ||
XM_021842733.1 | XP_021698425.1 | 4 - 5 | 5902 | 115 |

More Information
Set Membership: |
![]() ![]() |
