Assay Details
Gene Symbol: LOC5571158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844609.1 | XP_021700301.1 | 10 - 11 | 2388 | 96 | |
XM_021844611.1 | XP_021700303.1 | 11 - 12 | 2024 | 96 | ||
XM_021844612.1 | XP_021700304.1 | 11 - 12 | 1888 | 96 | ||
XM_021844613.1 | XP_021700305.1 | 9 - 10 | 2341 | 96 | ||
XM_021844614.1 | XP_021700306.1 | 10 - 11 | 2387 | 96 | ||
XM_021844615.1 | XP_021700307.1 | 9 - 10 | 1698 | 96 | ||
XM_021844616.1 | XP_021700308.1 | 9 - 10 | 2282 | 96 | ||
XM_021844617.1 | XP_021700309.1 | 9 - 10 | 1441 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
