Assay Details
Gene Symbol: LOC5577014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846935.1 | XP_021702627.1 | 8 - 9 | 3021 | 67 | |
XM_021846936.1 | XP_021702628.1 | 8 - 9 | 2953 | 67 | ||
XM_021846937.1 | XP_021702629.1 | 8 - 9 | 3006 | 67 | ||
XM_021846939.1 | XP_021702631.1 | 9 - 10 | 2880 | 67 | ||
XM_021846940.1 | XP_021702632.1 | 9 - 10 | 2879 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
