Assay Details
Gene Symbol: LOC5577014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846935.1 | XP_021702627.1 | 10 - 11 | 3342 | 71 | |
XM_021846936.1 | XP_021702628.1 | 10 - 11 | 3274 | 71 | ||
XM_021846937.1 | XP_021702629.1 | 10 - 11 | 3327 | 71 | ||
XM_021846939.1 | XP_021702631.1 | 11 - 12 | 3201 | 71 | ||
XM_021846940.1 | XP_021702632.1 | 11 - 12 | 3200 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
