Assay Details
Gene Symbol: LOC23687986 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011495273.2 | XP_011493575.2 | 3 - 4 | 485 | 87 | |
XM_021839925.1 | XP_021695617.1 | 1 - 2 | 472 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
