Assay Details
Gene Symbol: LOC5566523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845427.1 | XP_021701119.1 | 23 - 24 | 3476 | 56 | |
XM_021845429.1 | XP_021701121.1 | 23 - 24 | 3486 | 56 | ||
XM_021845430.1 | XP_021701122.1 | 23 - 24 | 3395 | 56 | ||
XM_021845431.1 | XP_021701123.1 | 23 - 24 | 3462 | 56 | ||
XM_021845432.1 | XP_021701124.1 | 23 - 24 | 3291 | 56 | ||
XM_021845433.1 | XP_021701125.1 | 23 - 24 | 3405 | 56 | ||
XM_021845434.1 | XP_021701126.1 | 23 - 24 | 3381 | 56 | ||
XM_021845435.1 | XP_021701127.1 | 23 - 24 | 3344 | 56 | ||
XM_021845436.1 | XP_021701128.1 | 23 - 24 | 3301 | 56 | ||
XM_021845437.1 | XP_021701129.1 | 23 - 24 | 3277 | 56 | ||
XM_021845438.1 | XP_021701130.1 | 23 - 24 | 3411 | 56 | ||
XM_021845440.1 | XP_021701132.1 | 23 - 24 | 3330 | 56 | ||
XM_021845441.1 | XP_021701133.1 | 23 - 24 | 3226 | 56 | ||
XM_021845442.1 | XP_021701134.1 | 23 - 24 | 3293 | 56 | ||
XM_021845443.1 | XP_021701135.1 | 22 - 23 | 3278 | 56 | ||
XM_021845444.1 | XP_021701136.1 | 23 - 24 | 3108 | 56 | ||
XM_021845445.1 | XP_021701137.1 | 21 - 22 | 2889 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
