Assay Details
Gene Symbol: LOC5566523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845427.1 | XP_021701119.1 | 3 - 4 | 1085 | 66 | |
XM_021845429.1 | XP_021701121.1 | 3 - 4 | 1095 | 66 | ||
XM_021845430.1 | XP_021701122.1 | 3 - 4 | 1004 | 66 | ||
XM_021845431.1 | XP_021701123.1 | 3 - 4 | 1071 | 66 | ||
XM_021845432.1 | XP_021701124.1 | 3 - 4 | 900 | 66 | ||
XM_021845433.1 | XP_021701125.1 | 3 - 4 | 1014 | 66 | ||
XM_021845434.1 | XP_021701126.1 | 3 - 4 | 990 | 66 | ||
XM_021845435.1 | XP_021701127.1 | 3 - 4 | 953 | 66 | ||
XM_021845436.1 | XP_021701128.1 | 3 - 4 | 910 | 66 | ||
XM_021845437.1 | XP_021701129.1 | 3 - 4 | 886 | 66 | ||
XM_021845438.1 | XP_021701130.1 | 3 - 4 | 1020 | 66 | ||
XM_021845440.1 | XP_021701132.1 | 3 - 4 | 939 | 66 | ||
XM_021845441.1 | XP_021701133.1 | 3 - 4 | 835 | 66 | ||
XM_021845442.1 | XP_021701134.1 | 3 - 4 | 902 | 66 | ||
XM_021845444.1 | XP_021701136.1 | 3 - 4 | 717 | 66 | ||
XM_021845446.1 | XP_021701138.1 | 3 - 4 | 1085 | 66 | ||
XM_021845447.1 | XP_021701139.1 | 3 - 4 | 1085 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021845427.1, XM_021845429.1, XM_021845430.1, XM_021845431.1, XM_021845432.1, XM_021845433.1, XM_021845434.1, XM_021845435.1, XM_021845436.1, XM_021845437.1, XM_021845438.1, XM_021845440.1, XM_021845441.1, XM_021845442.1, XM_021845444.1, XM_021845446.1, XM_021845447.1 transcripts.
Set Membership: |
![]() ![]() |
