Assay Details
Gene Symbol: LOC5566523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845427.1 | XP_021701119.1 | 2 - 3 | 889 | 69 | |
XM_021845430.1 | XP_021701122.1 | 2 - 3 | 808 | 69 | ||
XM_021845432.1 | XP_021701124.1 | 2 - 3 | 704 | 69 | ||
XM_021845435.1 | XP_021701127.1 | 2 - 3 | 757 | 69 | ||
XM_021845446.1 | XP_021701138.1 | 2 - 3 | 889 | 69 | ||
XM_021845447.1 | XP_021701139.1 | 2 - 3 | 889 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021845427.1, XM_021845430.1, XM_021845432.1, XM_021845435.1, XM_021845446.1, XM_021845447.1 transcripts.
Set Membership: |
![]() ![]() |
