Assay Details
Gene Symbol: LOC5568336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839258.1 | XP_021694950.1 | 6 - 7 | 934 | 82 | |
XM_021839259.1 | XP_021694951.1 | 7 - 8 | 1671 | 82 | ||
XM_021839260.1 | XP_021694952.1 | 5 - 6 | 658 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
