Assay Details
Gene Symbol: LOC5569559 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839490.1 | XP_021695182.1 | 9 - 10 | 3330 | 90 | |
XM_021839496.1 | XP_021695188.1 | 8 - 9 | 2428 | 90 | ||
XM_021839502.1 | XP_021695194.1 | 9 - 10 | 3330 | 90 | ||
XM_021839508.1 | XP_021695200.1 | 9 - 10 | 3330 | 90 | ||
XM_021839511.1 | XP_021695203.1 | 9 - 10 | 3330 | 90 | ||
XM_021839515.1 | XP_021695207.1 | 9 - 10 | 3330 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
