Assay Details
Gene Symbol: LOC5577693 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851987.1 | XP_021707679.1 | 2 - 3 | 766 | 111 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021851987.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
