Assay Details
Gene Symbol: LOC5578401 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841951.1 | XP_021697643.1 | 2 - 3 | 612 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5578401 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841951.1 | XP_021697643.1 | 2 - 3 | 612 | 64 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |