Assay Details
Gene Symbol: LOC5566871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854764.1 | XP_021710456.1 | 12 - 13 | 5041 | 65 | |
XM_021854765.1 | XP_021710457.1 | 11 - 12 | 5015 | 65 | ||
XM_021854766.1 | XP_021710458.1 | 12 - 13 | 4998 | 65 | ||
XM_021854767.1 | XP_021710459.1 | 12 - 13 | 5089 | 65 | ||
XM_021854768.1 | XP_021710460.1 | 11 - 12 | 4972 | 65 | ||
XM_021854769.1 | XP_021710461.1 | 11 - 12 | 5063 | 65 | ||
XM_021854770.1 | XP_021710462.1 | 12 - 13 | 4795 | 65 | ||
XM_021854771.1 | XP_021710463.1 | 12 - 13 | 4861 | 65 | ||
XM_021854772.1 | XP_021710464.1 | 11 - 12 | 4762 | 65 | ||
XM_021854773.1 | XP_021710465.1 | 11 - 12 | 4837 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
