Assay Details
Gene Symbol: LOC5566871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854764.1 | XP_021710456.1 | 10 - 11 | 4688 | 100 | |
XM_021854765.1 | XP_021710457.1 | 9 - 10 | 4662 | 100 | ||
XM_021854766.1 | XP_021710458.1 | 10 - 11 | 4645 | 100 | ||
XM_021854767.1 | XP_021710459.1 | 10 - 11 | 4736 | 100 | ||
XM_021854768.1 | XP_021710460.1 | 9 - 10 | 4619 | 100 | ||
XM_021854769.1 | XP_021710461.1 | 9 - 10 | 4710 | 100 | ||
XM_021854770.1 | XP_021710462.1 | 10 - 11 | 4442 | 100 | ||
XM_021854771.1 | XP_021710463.1 | 10 - 11 | 4508 | 100 | ||
XM_021854772.1 | XP_021710464.1 | 9 - 10 | 4409 | 100 | ||
XM_021854773.1 | XP_021710465.1 | 9 - 10 | 4484 | 100 |
