Assay Details
Gene Symbol: LOC110674038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837330.1 | XP_021693022.1 | 14 - 15 | 4500 | 85 | |
XM_021837332.1 | XP_021693024.1 | 14 - 15 | 4542 | 85 | ||
XM_021837333.1 | XP_021693025.1 | 14 - 15 | 4633 | 85 | ||
XM_021837334.1 | XP_021693026.1 | 13 - 14 | 4881 | 85 | ||
XM_021837335.1 | XP_021693027.1 | 15 - 16 | 4539 | 85 |
