Assay Details
Gene Symbol: LOC110674038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837330.1 | XP_021693022.1 | 6 - 7 | 1637 | 106 | |
XM_021837332.1 | XP_021693024.1 | 6 - 7 | 1679 | 106 | ||
XM_021837333.1 | XP_021693025.1 | 6 - 7 | 1770 | 106 | ||
XM_021837334.1 | XP_021693026.1 | 5 - 6 | 2018 | 106 | ||
XM_021837335.1 | XP_021693027.1 | 7 - 8 | 1676 | 106 |

More Information
Set Membership: |
![]() ![]() |
