Assay Details
Gene Symbol: LOC5568820 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658136.2 | XP_001658186.1 | 3 - 4 | 1315 | 60 | |
XM_021845642.1 | XP_021701334.1 | 3 - 4 | 1315 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
