Assay Details
Gene Symbol: LOC5567617 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840496.1 | XP_021696188.1 | 12 - 13 | 3548 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5567617 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840496.1 | XP_021696188.1 | 12 - 13 | 3548 | 60 |
Set Membership: |
![]() ![]() ![]() ![]() |