Assay Details
Gene Symbol: LOC5567617 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840496.1 | XP_021696188.1 | 4 - 5 | 1620 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021840496.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
