Assay Details
Gene Symbol: LOC5571798 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854066.1 | XP_021709758.1 | 12 - 13 | 1836 | 65 | |
XM_021854182.1 | XP_021709874.1 | 12 - 13 | 1821 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
