Assay Details
Gene Symbol: LOC5571798 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854009.1 | XP_021709701.1 | 9 - 10 | 1126 | 84 | |
XM_021854066.1 | XP_021709758.1 | 9 - 10 | 1126 | 84 | ||
XM_021854254.1 | XP_021709946.1 | 8 - 9 | 1069 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
