Assay Details
Gene Symbol: LOC5572548 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842605.1 | XP_021698297.1 | 2 - 3 | 1623 | 92 | |
XM_021842613.1 | XP_021698305.1 | 2 - 3 | 482 | 92 |
