Assay Details
Gene Symbol: LOC5572548 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842541.1 | XP_021698233.1 | 4 - 5 | 1864 | 103 | |
XM_021842546.1 | XP_021698238.1 | 4 - 5 | 1685 | 103 | ||
XM_021842552.1 | XP_021698244.1 | 4 - 5 | 2038 | 103 | ||
XM_021842557.1 | XP_021698249.1 | 4 - 5 | 1705 | 103 | ||
XM_021842562.1 | XP_021698254.1 | 4 - 5 | 1571 | 103 | ||
XM_021842569.1 | XP_021698261.1 | 4 - 5 | 1514 | 103 | ||
XM_021842574.1 | XP_021698266.1 | 4 - 5 | 1510 | 103 | ||
XM_021842575.1 | XP_021698267.1 | 4 - 5 | 1466 | 103 | ||
XM_021842579.1 | XP_021698271.1 | 4 - 5 | 1021 | 103 | ||
XM_021842584.1 | XP_021698276.1 | 4 - 5 | 842 | 103 | ||
XM_021842588.1 | XP_021698280.1 | 4 - 5 | 722 | 103 | ||
XM_021842594.1 | XP_021698286.1 | 4 - 5 | 720 | 103 | ||
XM_021842605.1 | XP_021698297.1 | 4 - 5 | 1862 | 103 | ||
XM_021842613.1 | XP_021698305.1 | 4 - 5 | 721 | 103 |

More Information
Set Membership: |
![]() ![]() |
