Assay Details
Gene Symbol: LOC5572548 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842541.1 | XP_021698233.1 | 5 - 6 | 1970 | 87 | |
XM_021842546.1 | XP_021698238.1 | 5 - 6 | 1791 | 87 | ||
XM_021842552.1 | XP_021698244.1 | 5 - 6 | 2144 | 87 | ||
XM_021842557.1 | XP_021698249.1 | 5 - 6 | 1811 | 87 | ||
XM_021842562.1 | XP_021698254.1 | 5 - 6 | 1677 | 87 | ||
XM_021842569.1 | XP_021698261.1 | 5 - 6 | 1620 | 87 | ||
XM_021842574.1 | XP_021698266.1 | 5 - 6 | 1616 | 87 | ||
XM_021842575.1 | XP_021698267.1 | 5 - 6 | 1572 | 87 | ||
XM_021842579.1 | XP_021698271.1 | 5 - 6 | 1127 | 87 | ||
XM_021842584.1 | XP_021698276.1 | 5 - 6 | 948 | 87 | ||
XM_021842588.1 | XP_021698280.1 | 5 - 6 | 828 | 87 | ||
XM_021842594.1 | XP_021698286.1 | 5 - 6 | 826 | 87 | ||
XM_021842605.1 | XP_021698297.1 | 5 - 6 | 1968 | 87 | ||
XM_021842613.1 | XP_021698305.1 | 5 - 6 | 827 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
